Aluminium ≥99.99%, folie, annealed, Thickness: 2.0 mm (0.08 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
40763.FI 40763.GH 40763.HP
40763.FIEA 2025 SEK
40763.FI 40763.GH 40763.HP
Aluminium ≥99.99%, folie, annealed, Thickness: 2.0 mm (0.08 in)
Aluminium
Purity: 99.99% CAS No.: 7429-90-5 Physical Form: ≈13.5g/50x50mm, Temper: soft Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321