Aluminium ≥99.999%, tråd, Puratronic®, Ø 0.18 mm (0.007 in)
Leverantör: Thermo Fisher Scientific
Synonyms: Aluminum
39730.G5 39730.G9
39730.G5EA 2500 SEK
39730.G5 39730.G9
Aluminium ≥99.999%, tråd, Puratronic®, Ø 0.18 mm (0.007 in)
Aluminium
Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈0.068g/m Odour: Odourless MDL No.: MFCD00134029
Formel: Al
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321