Aluminium ≥99.999%, stavar, Puratronic®, Ø 9.5 mm (0.37 in)
Leverantör: Thermo Scientific
Synonyms: Aluminum
10779.BQ 10779.BU 10779.CZ
10779.BQEA 2125 SEK
10779.BQ 10779.BU 10779.CZ
Aluminium ≥99.999%, stavar, Puratronic®, Ø 9.5 mm (0.37 in)
Aluminium
Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈1.91g/cm Odour: Odourless MDL No.: MFCD00134029
Formel: Al
MW: 26,98 g/mol
Kokpunkt: 2327 °C (1013 hPa)
Smältpunkt: 660 °C
Densitet: 2,7 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00134029
CAS nummer: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321