Magnesium ≥99.9%, folie, hard, Thickness: 0.125 mm (0.005 in)
Leverantör: Thermo Fisher Scientific
Fara
40602.FF 40602.FI
40602.FFEA 5950 SEK
40602.FF 40602.FI
Magnesium ≥99.9%, folie, hard, Thickness: 0.125 mm (0.005 in)
Magnesium
Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈0.14g/25x25mm, Temper: as rolled Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308
Formel: Mg
MW: 24.3 g/mol
Kokpunkt: 1107
Smältpunkt: 651
Storage Temperature: Omgivande
MDL Number: MFCD00085308
CAS nummer: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675