Magnesium, ribbon, Width: 3.0 mm (0.12 in), Thickness: 0.15 mm (0.0059 in)
Leverantör: Thermo Fisher Scientific
Fara
12811.15 12811.A0 12811.14 12811.51
12811.15EA 0 SEK
12811.15 12811.A0 12811.14 12811.51
Magnesium, ribbon, Width: 3.0 mm (0.12 in), Thickness: 0.15 mm (0.0059 in)
Magnesium
CAS No.: 7439-95-4 Physical Form: ≈0.78g/m Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308
Formel: Mg
Storage Temperature: Omgivande
MDL Number: MFCD00085308
CAS nummer: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III