Rifapentine 95%
Leverantör: Thermo Scientific
Varning
Synonyms: (2S,12Z,14E,16S,17S,18R,19R,20R,21S,22R,23S,24E)-3-(((4-Cyclopentyl-1-piperazinyl)imino)methyl)-21-(Acetyloxy)-5,6,9,17,19-pentahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-2,7-(epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-1,11(2H)-dione
461900250 461901000
ACRO461900250EA 1700 SEK
ACRO461900250 ACRO461901000
Rifapentine 95%
Rifapentine
Formel: C₄₇H₆₄N₄O₁₂
MW: 877,04 g/mol
Smältpunkt: 179…180 °C
Storage Temperature: Frys
MDL Number: MFCD00866806
CAS nummer: 61379-65-5
Order Now
Specifikation testresultat
Appearance Red to dark red Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤8 %
HPLC ≥94 %