Nicotinamide-N-oxide 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Nicotinamide N-oxide, Pyridine-3-carboxamide N-oxide, 3-Carbamoylpyridine 1-oxide
H61292.14 H61292.22
H61292.14EA 5575 SEK
H61292.14 H61292.22
Nicotinamide-N-oxide 98%
Nicotinamide-N-oxide
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Smältpunkt: 291…293 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006202
CAS nummer: 1986-81-8
EINECS: 217-859-4
Order Now
Specifikation testresultat
Assay 97,5%