Penicillin V potassium salt 99%
Leverantör: Thermo Scientific
Fara
Synonyms: Potassium (2S,5R,6R)-3,3-dimethyl-7-oxo-6-(2-phenoxyacetamido)-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate, Penicillin V Potassium
455230250 455230050
ACRO455230250EA 1900 SEK
ACRO455230250 ACRO455230050
Penicillin V potassium salt 99%
Penicillin V potassium salt
Formel: C₁₆H₁₇KN₂O₅S
MW: 388,48 g/mol
Smältpunkt: 197…202 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00051771
CAS nummer: 132-98-9
Order Now
Specifikation testresultat
Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +220° to +235° (20°C, 589 nm) (c=1, H2O)