6-tert-Butyl-2,4-xylenol 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 2-(tert-Butyl)-4,6-dimethylphenol
462570100 462570500
ACRO462570100EA 850 SEK
ACRO462570100 ACRO462570500
6-tert-Butyl-2,4-xylenol 97%
6-tert-Butyl-2,4-xylenol
Formel: C₁₂H₁₈O
MW: 178,28 g/mol
Kokpunkt: 249 °C
Smältpunkt: 22…23 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002234
CAS nummer: 1879-09-0
UN: 2811
ADR: 6.1,I
Order Now
Specifikation testresultat
Appearance White to light yellow Liquid or solid
Infrared spectrum Conforms
GC ≥96.0 %