Ethynylestradiol 98%
Leverantör: Thermo Fisher Scientific
Fara
461550010 461550050
ACRO461550010EA 2025 SEK
ACRO461550010 ACRO461550050
Ethynylestradiol 98%
Ethynylestradiol
Formel: C₂₀H₂₄O₂
Smältpunkt: 180…188 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00003690
CAS nummer: 57-63-6
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 181 to 185 °C
HPLC ≥97.5 %