Prasugrel 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 5-[2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl]-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl Acetate, 5-(2-Cyclopropyl-1-(2-fluorophenyl)-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridin-2-yl acetate
461480010 461482500
ACRO461480010EA 2725 SEK
ACRO461480010 ACRO461482500
Prasugrel 98%
Prasugrel
Formel: C₂₀H₂₀FNO₃S
MW: 373,44 g/mol
Smältpunkt: 110…121 °C
Storage Temperature: Frys
MDL Number: MFCD09954140
CAS nummer: 150322-43-3
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %