2-Methylpyrimidine 98%
Leverantör: Thermo Fisher Scientific
Fara
459860010 459860050
ACRO459860010EA 1700 SEK
ACRO459860010 ACRO459860050
2-Methylpyrimidine 98%
2-Methylpyrimidine
Formel: C₅H₆N₂
Kokpunkt: 130…132 °C (760 mmHg)
Smältpunkt: –6 °C
Densitet: 1,0132 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00128182
CAS nummer: 5053-43-0
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4940 (20°C, 589 nm)
Water ≤2.5 %
GC ≥97.5 %