Natriumdodecylsulfat, granular, pure, meets analytical specification of Ph.Eur.
Leverantör: Thermo Fisher Scientific
Fara
459000010 459000050
ACRO459000010EA 1050 SEK
ACRO459000010 ACRO459000050
Natriumdodecylsulfat, granular, pure, meets analytical specification of Ph.Eur.
Natriumdodecylsulfat
Formel: CH₃(CH₂)₁₁OSO₃Na
Smältpunkt: 206 °C
Flampunkt: >150 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036175
CAS nummer: 151-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or crystals
Identification A Passes test
Identification B Passes test
Identification C Passes test
Identification D Passes test
Alkalinity Passes test
Titration ≥85 %
NaCl and Na2SO4 ≤8 %
Non-esterified alcohols ≤4 %