Propionylklorid 99.5% phosgene-free
Leverantör: Thermo Scientific
Fara
Synonyms: Propanoylklorid
458970250 458975000
458970250.EA 430 SEK
458970250. 458975000.
Propionylklorid 99.5% phosgene-free
Propionylklorid
Formel: C₃H₅ClO
MW: 92,52 g/mol
Kokpunkt: 80 °C (760 mmHg)
Smältpunkt: –94 °C
Densitet: 1,059 g/cm³
Flampunkt: 6 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000745
CAS nummer: 79-03-8
UN: 1815
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4020 to 1.4050 (20°C, 589 nm)
GC ≥99.4 %
Color scale ≤20 APHA
Titration RCOCl ≥99.4 %