Jod, meets analytical specification of BP, Ph. Eur., USP
Leverantör: Thermo Fisher Scientific
Fara
457760010 457760500 457762500
ACRO457760010EA 4150 SEK
ACRO457760010 ACRO457760500 ACRO457762500
Jod, meets analytical specification of BP, Ph. Eur., USP
Jod
Formel: I₂
Kokpunkt: 185 °C (760 mmHg)
Smältpunkt: 113 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011355
CAS nummer: 7553-56-2
UN: 3495
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Greyish-violet Brittle plates or crystals with metallic sheen
Titration Iodometric 99.8 to 100.5 %
Non-volatile matter ≤0.05 %
Chloride and Bromide ≤250 ppm