5-Metylhexansyra 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 5-Methylhexanoic acid, 5-Methylhexanoicacid
457410050 457410250 457411000
ACRO457410050EA 585 SEK
ACRO457410050 ACRO457410250 ACRO457411000
5-Metylhexansyra 98%
5-Metylhexansyra
Formel: C₇H₁₄O₂
MW: 130,19 g/mol
Kokpunkt: 121 °C (30 mmHg)
Densitet: 0,905 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00036494
CAS nummer: 628-46-6
UN: 3265
ADR: 8,III
Order Now
Specifikation testresultat
Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥97.5 %