2,3,6,7,10,11-Hexahydroxytriphenylene 95%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Triphenylene-2,3,6,7,10,11-hexaol
457010010 457010050
ACRO457010010EA 1725 SEK
ACRO457010010 ACRO457010050
2,3,6,7,10,11-Hexahydroxytriphenylene 95%
2,3,6,7,10,11-Hexahydroxytriphenylene
Formel: C₁₈H₁₂O₆
MW: 324.29 g/mol
Smältpunkt: 138 °C
Storage Temperature: Omgivande
MDL Number: MFCD01321170
CAS nummer: 4877-80-9
Order Now
Specifikation testresultat
Appearance Grey to purple-grey Powder
Infrared spectrum Conforms
HPLC ≥94.0 %
NMR (1H-NMR) Consistent with structure