Tazobactam sodium
Leverantör: Thermo Scientific
455370010
455372500
ACRO455370010EA
4375
SEK
ACRO455370010
ACRO455372500
Tazobactam sodium
Tazobactam sodium
Formel:
C₁₀H₁₁N₄NaO₅S Smältpunkt: 140…147 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00917472 CAS nummer: 89785-84-2 |
Specifikation testresultat
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤10 % |
HPLC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...