Ethyl viologen dibromide 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 1,1'-Diethyl-[4,4'-bipyridine]-1,1'-diium bromide, 1,1'-Diethyl-4,4'-bipyridinium dibromide
455030010 455030050
ACRO455030010EA 650 SEK
ACRO455030010 ACRO455030050
Ethyl viologen dibromide 97%
Ethyl viologen dibromide
Formel: C₁₄H₁₈Br₂N₂
MW: 374.12 g/mol
Smältpunkt: 278 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00134590
CAS nummer: 53721-12-3
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %