Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%
Leverantör: Thermo Scientific
Varning
Synonyms: TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine
454531000 454535000
ACRO454531000EA 3275 SEK
ACRO454531000 ACRO454535000
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Formel: C₃₀H₃₀N₁₀
MW: 530,64 g/mol
Smältpunkt: 138…140 °C
Storage Temperature: Omgivande
MDL Number: MFCD09265124
CAS nummer: 510758-28-8
Order Now
Specifikation testresultat
Appearance White to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Elemental Analysis 25.34 to 27.45 % (N)