4-[1,2,4]Triazol-4-yl-benzoic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-(4H-1,2,4-Triazol-4-yl)benzoic acid, 4-(p-Carboxyphenyl)-1,2,4-triazole
449960010 449960050
ACRO449960010EA 4675 SEK
ACRO449960010 ACRO449960050
4-[1,2,4]Triazol-4-yl-benzoic acid 97%
4-[1,2,4]Triazol-4-ylbenzoic acid
Formel: C₉H₇N₃O₂
MW: 189.18 g/mol
Smältpunkt: 330 °C
Storage Temperature: Omgivande
MDL Number: MFCD06654839
CAS nummer: 157069-48-2
Order Now
Specifikation testresultat
Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %