Sodium methanesulphonate 99%
Leverantör: Thermo Scientific
Synonyms: Methanesulphonic acid sodium salt
442110050 442111000 442115000
ACRO442110050EA 650 SEK
ACRO442110050 ACRO442111000 ACRO442115000
Sodium methanesulphonate 99%
Sodium methanesulphonate
Formel: CH₃NaO₃S
MW: 118,09 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00064389
CAS nummer: 2386-57-4
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
Titration after Ion exchange ≥98.5 %
Free acid ≤1 %