(±)-γ-Hexalactone 98%
Leverantör: Thermo Scientific
Synonyms: (±)-γ-Caprolactone, γ-Caprolactone, 5-Ethyldihydrofuran-2(3H)-one, (±)-Hexan-4-olide
394170250 394171000 394175000
ACRO394170250EA 395 SEK
ACRO394170250 ACRO394171000 ACRO394175000
(±)-γ-Hexalactone 98%
(±)-γ-Hexalactone
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Kokpunkt: 220 °C (760 mmHg)
Smältpunkt: –18 °C
Densitet: 1,023 g/cm³
Flampunkt: 98 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005401
CAS nummer: 695-06-7
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20°C, 589 nm)
GC ≥97.5 %