5,6,7,8-Tetrahydroquinoline 98%
Leverantör: Thermo Scientific
Varning
188870050 188870250 188871000
ACRO188870050EA 770 SEK
ACRO188870050 ACRO188870250 ACRO188871000
5,6,7,8-Tetrahydroquinoline 98%
5,6,7,8-Tetrahydroquinoline
Formel: C₉H₁₁N
MW: 133,19 g/mol
Kokpunkt: 218…222 °C (760 mmHg)
Densitet: 1,03 g/cm³
Flampunkt: 86 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006734
CAS nummer: 10500-57-9
Order Now
Specifikation testresultat
Appearance Colorless to light brown Liquid
Infrared spectrum Conforms
GC ≥98.5 %