2-Cyclohepten-1-one 90%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Cyclohept-2-enone
171070010 171070050
ACRO171070010EA 1475 SEK
ACRO171070010 ACRO171070050
2-Cyclohepten-1-one 90%
2-Cyclohepten-1-one
Formel: C₇H₁₀O
MW: 110.16 g/mol
Kokpunkt: 189 °C (760 mmHg)
Densitet: 0.99 g/cm³
Flampunkt: 68 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004157
CAS nummer: 1121-66-0
Order Now
Specifikation testresultat
Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥89.0 %
HPLC ≥94 %