1,2,4,5-Tetrametylbensen 97+%
Leverantör: Thermo Scientific
Fara
138370050 138375000 138371000
ACRO138370050EA 555 SEK
ACRO138370050 ACRO138375000 ACRO138371000
1,2,4,5-Tetrametylbensen 97+%
1,2,4,5-Tetrametylbensen
Formel: C₁₀H₁₄
MW: 134,22 g/mol
Kokpunkt: 196…197 °C (760 mmHg)
Smältpunkt: 78…82 °C
Flampunkt: 73 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008528
CAS nummer: 95-93-2
UN: 1325
ADR: 4.1,II
Order Now
Specifikation testresultat
Appearance Semitransparent white to brown Crystals
Infrared spectrum Conforms
Melting point 78 to 82 °C
GC ≥97.0 %
Elemental Analysis 4.5 to 4.9 % N