Atropic acid 97%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Phenylacrylic Acid, Ipratropium bromide Impurity D
459050050 459050250
ACRO459050050EA 1200 SEK
ACRO459050050 ACRO459050250
Atropic acid 97%
Atropic acid
Formel: C₉H₈O₂
MW: 148,16 g/mol
Kokpunkt: 267 °C
Smältpunkt: 104…106 °C
Storage Temperature: Omgivande
MDL Number: MFCD00046531
CAS nummer: 492-38-6
Order Now
Specifikation testresultat
Appearance White to light yellow Powder or crystalline powder
Infrared spectrum Conforms
Melting point 104 to 110 °C
HPLC ≥96.0 %