3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 3,5-Di-tert-butylsalicylaldehyde
458120050 458120250
ACRO458120050EA 1025 SEK
ACRO458120050 ACRO458120250
3,5-Di-tert-butyl-2-hydroxybenzaldehyde 99%
3,5-Di-tert-butyl-2-hydroxybenzaldehyde
Formel: C₁₅H₂₂O₂
MW: 234,34 g/mol
Smältpunkt: 61…62 °C
Storage Temperature: Omgivande
MDL Number: MFCD00191998
CAS nummer: 37942-07-7
Order Now
Specifikation testresultat
Appearance White to yellow Powder or crystalline powder
Infrared spectrum Conforms
Melting point 56 to 63 °C
GC ≥98.5 %