Pentafluorobenzoic acid 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2,3,4,5,6-Pentafluorobenzoic acid
129910050 129910250 129911000
ACRO129910050EA 425 SEK
ACRO129910050 ACRO129910250 ACRO129911000
Pentafluorobenzoic acid 99%
Pentafluorobenzoic acid
Formel: C₇HF₅O₂
MW: 212,08 g/mol
Kokpunkt: 220 °C (760 mmHg)
Smältpunkt: 100…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002406
CAS nummer: 602-94-8
Order Now
Specifikation testresultat
Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 104 °C
GC ≥98.5 %