SRK® Surface Control Swabs
Leverantör: Copan
906C.CR 906C 9U002S 9U017S 9U011S01 9U012S
710-1976EA 9550 SEK
710-1976 710-1975 710-1977 710-2232 710-2231 COPA9U012S
SRK® Surface Control Swabs
Svabbar Svabbar, provtagning
Surface control swabs and sponges designed for the hygiene monitoring of surfaces.
  • High recovery for bioburden control
  • Enables validation of sanitization process
Collection and transport systems for microbiological applications in food, hospitals and other sectors such as cosmetics and industries.
Förpackning: VHP resistant triple-protection. Individually wrapped.