Isooktan, dehydrerad (max. 0,005% H2O)
Leverantör: VWR Chemicals
Fara
85821.230EA 900 SEK
85821.230 85821.269
Isooktan, dehydrerad (max. 0,005% H2O)
Isooktan
Formel: (CH₃)₂CHCH₂C(CH₃)₃
Kokpunkt: 99,2 °C (1013 hPa)
Smältpunkt: –107 °C
Densitet: 0,692 g/cm³ (20 °C)
Flampunkt: –12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008943
CAS nummer: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3,II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay (GC, on anhydrous basis) 99.5-100 %
Residue after evaporation 0-0.001 % w/w
Water (KF) 0-0.005 % w/w
Acidity (as Acetic acid) 0-0.003 %