1,2-Dikloretan, dehydrerad (max. 0,005% H2O)
Leverantör: VWR Chemicals
Fara
Synonyms: Etylendiklorid
85816.230EA 900 SEK
85816.230 85816.269
1,2-Dikloretan, dehydrerad (max. 0,005% H2O)
1,2-Dikloretan
250 and 500 ml bottles with a septum cap featuring six separate re-sealable puncture points
Formel: ClH₂CCH₂Cl
Kokpunkt: 83,5 °C (1013 hPa)
Smältpunkt: –35,5 °C
Densitet: 1,254 g/cm³ (20 °C)
Flampunkt: 13 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000963
CAS nummer: 107-06-2
EINECS: 203-458-1
UN: 1184
ADR: 3 (6.1),II
REACH: 01-2119484658-20
Merck Index: 13,03831
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48789e88e6041156e33712fd7c0198d7a4910266&vid=4238623c54017660b83d3baa856a69cc1aa8ee6d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay (GC, on anhydrous basis) 99.8-100 %
Acidity (as HCl) 0-0.001 %
Residue after evaporation 0-0.001 % w/w
Water (KF) 0-0.005 % w/w