Metyl-tert-butyleter, dehydrerad (max. 0,005% H2O)
Leverantör: VWR Chemicals
Fara
85814.230EA 0 SEK
85814.230 85814.269
Metyl-tert-butyleter, dehydrerad (max. 0,005% H2O)
Metyl-tert-butyleter
Formel: (CH₃)₃COCH₃
Kokpunkt: 55,2 °C (1013 hPa)
Densitet: 0,7405 g/cm³ (20 °C)
Flampunkt: −28 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008812
CAS nummer: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Assay (GC, on anhydrous basis) 99-100 %
Acidity 0-0.001 meq/g
Peroxides (as H202) 0-0.0005 %
Residue after evaporation 0-0.001 % w/w
Water (KF) 0-0.005 % w/w