Quantus® AreaDress, Cleanroom Trousers
Leverantör: VWR Collection
113-9187EA 0 SEK
113-9187 113-9186 113-9190 113-9189 113-9191 113-9192 113-9188
Quantus® AreaDress, Cleanroom Trousers
Workwear
These trousers are made from extremly breathable polyester fabric with conductive yarn and a PTFE finish that provides good splash protection against liquids.
  • Wide, double stitched comfort waistband
  • Knitted cuffs, no pockets
  • Excellent wearing comfort
  • Machine washable up to 60 °C
  • Not autoclavable
Recommended for use in controlled production environments.
Weight: 116 g/m²
Certifikat: Oeko-Tex® Standard 100 certified.