Fmoc-Lys(Z)-OH 98%
Leverantör: Thermo Fisher Scientific
Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysine, Fmoc-Lys(Cbz)-OH, Fmoc-Lys(Z)-OH, Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, Nalpha-Fmoc-Nepsilon-Cbz-L-lysine
H62866.06 H62866.14
H62866.06EA 2125 SEK
H62866.06 H62866.14
Fmoc-Lys(Z)-OH 98%
Fmoc-Lys(Z)-OH
Formel: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Smältpunkt: 110…120 °C
Storage Temperature: Frys
MDL Number: MFCD00065662
CAS nummer: 86060-82-4
Order Now
Specifikation testresultat
Purity 97,5%
Purity method Assay (HPLC)