Clelandreagens, SPUTOLYSIN®
Leverantör: Merck Millipore Calbiochem
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
560000-1 560000-10
560000-1EA 10250 SEK
560000-1 560000-10
Clelandreagens, SPUTOLYSIN®
Clelandreagens
Spotolysin reagent is a concentrated solution of Dithiothreitol in phosphate buffer. Mainly for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum.
  • Epethelial cell isolation
  • Concentrate of dithiothreitol (DTT) in phosphate buffer
  • Isolation from sputum
  • pH 6.5 - 7.5
Each vial is sufficient to make 100 ml of working solution.
Formel: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00004877
CAS nummer: 3483-12-3