Metyl-tert-butyleter, AR®, Laboratory Reagent, Macron Fine Chemicals™
Leverantör: Avantor Performance Materials
Synonyms: MTBE
5449-45EA 4425 SEK
5449-45 5449-25
Metyl-tert-butyleter, AR®, Laboratory Reagent, Macron Fine Chemicals™
Metyl-tert-butyleter
Formel: (CH₃)₃COCH₃
MW: 88.15 g/mol
Kokpunkt: 55.2
Smältpunkt: -108.6
Flampunkt: -28
MDL Number: MFCD00008812
CAS nummer: 1634-04-4
UN: 2398
ADR: 3 II
Merck Index: 13,06059
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1712bc714ba0a06e0de37f7cf4e5117256f84425&vid=e1b314f829136075329c50f9e5a593c19ae28b4f&prid=web&clid=SAPDAM