2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
Leverantör: Avantor Performance Materials
Synonyms: Isopropanol
6775-25EA 735 SEK
6775-25 6775-45 6775-39 MACR6775-16
2-Propanol, AR®, Laboratory Reagent, Macron Fine Chemicals™
2-Propanol
Formel: (CH₃)₂CHOH
MW: 60.1 g/mol
Kokpunkt: 82
Smältpunkt: -89
Flampunkt: 12
MDL Number: MFCD00011674
CAS nummer: 67-63-0
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM