Saltsyra 10% aqueous solution NF, Macron Fine Chemicals™
Leverantör: Avantor Performance Materials
Fara
Synonyms: Hydrochloric acid solution, Muriatic acid
2608-08EA 0 SEK
2608-08 2608-22 2608-45 2608-02
Saltsyra 10% aqueous solution NF, Macron Fine Chemicals™
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
Order Now
Specifikation testresultat
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 9.5 - 10.5
NF - Assay (HCl)(g/100ml) 9.5 - 10.5
NF - Identification (Cl) Passes Test
NF - Residue on Ignition ≤2 mg
NF - Free Bromine or Chlorine Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Sulfite (SO₃) Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.