Citronsyra monohydrat E330, Ph. Eur., Macron Fine Chemicals™
Leverantör: Avantor Performance Materials
Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
P101-33EA 0 SEK
P101-33 P101-38
Citronsyra monohydrat E330, Ph. Eur., Macron Fine Chemicals™
Citronsyra monohydrat
Formel: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Flampunkt: 173,9
MDL Number: MFCD00149972
CAS nummer: 5949-29-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM