Nourseothricin sulphate, Molecular Biology Grade
Leverantör: G Biosciences
Synonyms: NTC, Streptothricin sulphate, clonNAT
RC-188 RC-189 RC-190 RC-187
GENORC-188EA 3575 SEK
GENORC-188 GENORC-189 GENORC-190 GENORC-187
Nourseothricin sulphate, Molecular Biology Grade
Nourseothricin sulphate
Nourseothricin sulphate is a Streptothricin-class antibiotic. It has an extremely broad spectrum of activity against viruses, bacteria, eukaryotic unicellular and complex organisms. Norseothricin contains both Streptothricin D (C₃₁H₅₈N₁₂O₁₀) and Streptothricin F (C₁₉H₃₄N₈O₈) components.
Formel: C₃₁H₅₈N₁₂O₁₀·C₁₉H₃₄N₈O₈·H₂SO₄
MW: 1359,47 g/mol
MDL Number: MFCD00165013
CAS nummer: 96736-11-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=753248babe43663500b273b4c6f6b13baa87f079&vid=d53933f2563b93b50e2ff62374c33073fac9d775&prid=web&clid=SAPDAM