VWR®, Low Temperature PCR Rack, 96 Well
Leverantör: VWR Collection
732-2975EA 1225 SEK
732-2975 732-3319 732-3320 732-3321
VWR®, Low Temperature PCR Rack, 96 Well
Ställ Tube Racks
This PCR storage rack offers a sleek and modern design that includes a unique lid that the rack slides out from, and then rotates 180°, to become a convenient stand that the rack then sits on. Ergonomic, textured finger grips provide a secure hold, reducing the chance of dropping the PCR rack, making handling and transportation easy, especially when using gloves.
  • Holds 0,2 ml tubes, individual or strip
  • 96-well (8×12 configuration)
  • For freezer storage down to –80 °C
  • Detachable lid
The lid has a side opening that enables faster cooling of samples to ensure integrity, and is made from translucent polycarbonate for easy sample and well identification. The PCR rack base is made from robust ABS construction that helps keep samples safe, so you don’t have to spend extra money and time on expensive re-testing. A location ID of numbers and letters allows for organised sample placement, which results in easy and quick sample retrieval. It has a closed base design that helps maintain sample integrity by staying cold for longer and is versatile enough to be used on a bench top or for freezer storage. Stackable, which helps save valuable space in the laboratory, and available in multiple colours for coding options. The lid is ambidextrous; it can be switched to accommodate left or right hand orientation.
Assorted colours (blue, purple, green, orange and yellow).