Calorimetric standards
Leverantör: PARR
3415 3416 3417
PARR3415EA 3600 SEK
PARR3415 PARR3416 PARR3417
Calorimetric standards
Standarder Standarder för miljöanalys
Calorific standard in powder or pellet form for standardising all oxygen bomb calorimeters.