Acetonitril s 0,1% (v/v) Myrsyra, LiChrosolv®, hypergrade for LC-MS, Supelco®
Leverantör: Merck
Fara
M-Clarity™ quality level = MQ100
1.59002.1000 1.59002.2500 1.59002.4000
1.59002.1000EA 1290 SEK
1.59002.1000 1.59002.2500 1.59002.4000
Acetonitril s 0,1% (v/v) Myrsyra, LiChrosolv®, hypergrade for LC-MS, Supelco®
Acetonitril s 0,1% Myrsyra
LiChrosolv® hypergrade is accurately tested for LC-MS suitability, meeting all the requirements of modern LC-MS ionisation methods (ESI / APCI - positive and negative mode).
Thanks to their low level of ionic background and low ion suppression, LiChrosolv® hypergrade solvents provide high reproducibility and high ionisation efficiency. The packaging material has been improved to prefectly meet LC-MS quality requirements.
Kokpunkt: 81,6 °C (1013 hPa)
Flampunkt: 2 °C
Storage Temperature: Omgivande
UN: 1648
ADR: 3,