Safflower oil for food industry
Leverantör: MP Biomedicals
0210288890 0210288891
ICNA0210288890EA 1500 SEK
ICNA0210288890 ICNA0210288891
Safflower oil for food industry
Safflower oil
Storage Temperature: Omgivande
MDL Number: MFCD00132216
CAS nummer: 8001-23-8