Real-Time PCR kit for detection of toxic Vibrio cholerae
Leverantör: Q BIOANALYTIC
QB-RTI-19-1000 QBRTI-19-10SMP QB-RTI-19-250 QB-RTI-19-50
710-1765EA 0 SEK
710-1765 710-1766 710-1767 710-1768
Real-Time PCR kit for detection of toxic Vibrio cholerae
Testsystem för mikrobiologi PCR Tests
Kit for Vibrio cholerae tox gene (ctx). Food safety experts like the German DGHM recommend to test warm water fish for vibrio cholerae. With this Real-Time PCR test you have a very versatile method for testing whether or not the present strain has the ability to produce toxins. You can use our Real-Time PCR Test for Vibrio cholerae for testing for the presence of the species before. You can use this test in combination with an appropriate pre- enrichment in alkaline peptone-water or you can confirm bacterial colonies on agar plates.
  • TaqMan probes
  • Contains internal amplification control in HEX channel
  • Taget in FAM channel
  • Contains UNG to protect from re-amplification
  • Positive DNA and negative control included
Certifikat: DIN EN ISO 20838 conform detection in food, feed and water samples from pre-enrichment cultures or for confirmation of bacterial colonies.