Lupeol ≥98%, kristaller
Leverantör: Cayman Chemical
Varning
Synonyms: Fagarasterol, Monogynol B, Clerodol, 20(29)-Lupen-3β-ol, 3β-Hydroxy-20(29)-lupene
11215-5 11215-10 11215-25
CAYM11215-5EA 935 SEK
CAYM11215-5 CAYM11215-10 CAYM11215-25
Lupeol ≥98%, kristaller
Lupeol
Lupeol, a dietary triterpene found in certain fruits, vegetables, and medicinal plants, has potent anti-inflammatory, anticarcinogenic, antimutagenic, and antimalarial activity. It suppresses the growth of hepatocellular carcinoma cell lines SMMC7721 and HepG2 with IC50 values of 45 and 48.5 μM and melanoma cell lines Mel 928 and Mel 1241 with IC50 values of 75 and 72 μM.¹'² At 0.76 g/kg lupeol causes a significant decrease in the blood pressure of stroke-prone hypertensive rats and reduces expression of hepatic genes involved in triglyceride and cholesterol synthesis.³ It also has significant anti-inflammatory effects at 50 mg/kg in a carrageenan-induced edema model, inhibiting neutrophil migration.⁴
1. Tarapore, R.S.,Siddiqui, I.A.,Saleem, M., et al. Specific targeting of Wnt/β-catenin signaling in human melanoma cells by a dietary triterpene lupeol. Carcinogenesis 31(10), 1844-1853 (2010).
2. He, Y.,Liu, F.,Zhang, L., et al. Growth inhibition and apoptosis induced by lupeol, a dietary triterpene, in human hepatocellular carcinoma cells. Bio.Pharm.Bull. 34(4), 517-522 (2011).
3. Ardiansyah,Yamaguchi, E.,Shirakawa, H., et al. Lupeol supplementation improves blood pressure and lipid metabolism parameters in stroke-prone spontaneously hypertensive rats. Bioscience, Biotechnology, and Biochemistry 76(1), 183-185 (2012).
4. Lucetti, D.L.,Lucetti, E.C.,Bandeira, M.A., et al. Anti-inflammatory effects and possible mechanism of action of lupeol acetate isolated from Himatanthus drasticus (Mart.) Plumel. Journal of Inflammation 7(60), 1-11 (2010).
Formel: C₃₀H₅₀O
MW: 426,7 g/mol
Storage Temperature: Frys
CAS nummer: 545-47-1