Etanol absolut VLSI
Leverantör: VWR Chemicals
Fara
Synonyms: Ethyl alcohol
85651.320EA 470 SEK
85651.320 85651.360
Etanol absolut VLSI
Etanol
Observera: WARNING : This alcohol is regulated. Please contact VWR.
Formel: C₂H₅OH
MW: 46.07 g/mol
Kokpunkt: 78.3 °C (1013 hPa)
Smältpunkt: –117 °C
Densitet: 0.7895 g/cm³ (20 °C)
Flampunkt: 12 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00003568
CAS nummer: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806