Metanol VLSI
Leverantör: VWR Chemicals
Fara
Synonyms: Hydroxy methane, Carbinol, Wood alcohol, Methyl alcohol
85650.320EA 420 SEK
85650.320 85650.360
Metanol VLSI
Metanol
Formel: CH₃OH
MW: 32,04 g/mol
Kokpunkt: 64,6 °C (1013 hPa)
Smältpunkt: –98 °C
Densitet: 0,7918 g/cm³ (20 °C)
Flampunkt: 11 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00004595
CAS nummer: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=2d58b015073a1ab1f0ee9e0aa563ea1c28de4d5e&prid=web&clid=SAPDAM