Anti-CALB1 Mouse Monoclonal Antibody [clone: CL-300]

Leverantör: DIAGNOSTIC BIOSYSTEM

MOB295-05 MOB295
DIABMOB295-05EA 0 SEK
DIABMOB295-05 DIABMOB295
Anti-CALB1 Mouse Monoclonal Antibody [clone: CL-300]
Type:
Antigen: CALB1
Clonality: Monoclonal
Clone: CL-300
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR